Description
The clamping ring is a component and therefore a spare part for the KECK™ Adapter KA from glass olive to plastic olive. It is available for threads GL 14 and RD 14.
- The clamping ring is a component of the KECK™ Adapter KA and connects the glass olive with the plastic olive.
- available for threads GL 14 and RD 14
Specifications
Specifications
| Quantity | 100 |
| For Use With (Equipment) | Sealing |
| For Use With (Application) | Sealing |
| Size | GL 14 |
| Color | Yellow |