Learn More
Lenz Laborglasinstrumente™ Keck™ Conical Joint Clips - Nickel
Description
Pack of 10
Specifications
Specifications
| Quantity | 10 |
| Material | Nickel |
| For Use With (Equipment) | For ground joints NS 19/26 |