Learn More
DWK Life Sciences™ KECK™ Clip for Conical Joints, POM, KC
Product Code. 10468762 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
NS 10
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 40
NS 45
Color:
Amber
Blue
Green
Orange
Purple
Red
Yellow
Unit Size:
Each
Pack of 100
Product Code. Size Color unitSize
10468762 NS 34 Orange Each
10039920 NS 10 Green Each
15884678 NS 14 Yellow Pack of 100
15894678 NS 19 Blue Pack of 100
15804688 NS 24 Green Pack of 100
15814688 NS 29 Red Pack of 100
15874678 NS 12 Purple Pack of 100
15981571 NS 40 Yellow Pack of 100
10633982 NS 45 Amber Pack of 100
9 options
Product Code. 10468762
Brand: DWK Life Sciences™ 286336709
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
KECK™ conical ground clamps for a safe ground joint.
KECK™ POM (polyoxymethylene) conical ground joint clamps are easy, safe and quick to attach and are characterized by their dimensional stability and good spring properties. In contrast to conventional wire clamps, the glass surface is not damaged when it is attached. This increases safety in the laboratory. The sizes can be easily distinguished from each other by the different colours.
  • Excellent dimensional stability and good spring properties
  • No damage to the glass surface during attachment unlike conventional wire clamps
  • Easy, safe and quick to put on and take off
  • Colour coding per size
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100
Material POM
For Use With (Equipment) Securing of Conical Ground Joints
For Use With (Application) Securing of conical ground joints
Size NS 34
Color Orange