Learn More
DWK Life Sciences™ KECK™ Clip, for conical joints, from POM
Product Code. 10749474 Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 40
NS 45
Color:
Amber
Blue
Green
Orange
Red
Violet
Yellow
Unit Size:
Pack of 10
Product Code. 10749474
Brand: DWK Life Sciences™ 1091148
Only null left
Add to basket
Add to basket
The KECK™ clamp is used to secure a standard ground joint connection.
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
  • For conical joints
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
  • Increased safety in the laboratory as the glass surface is not scratched
  • Marking of the individual sizes for better differentiation
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10
Material POM
For Use With (Equipment) Secure Glass-to-Glass Joints
For Use With (Application) Secure glass-to-glass joints
Size NS 34
Color Orange