Learn More
DWK Life Sciences™ KECK™ Clip for Conical Joints, Ni, KCM
Product Code. 31781798 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
NS 14
NS 19
NS 24
NS 29
Unit Size:
Each
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31781798 NS 19 Pack of 100
10579181 NS 14 Each
15824688 NS 29 Pack of 100
10394911 NS 19 Each
15901581 NS 24 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31781798 Supplier DWK Life Sciences™ Supplier No. 286337405
KECK™ conical ground joint clamps for high temperature applications.
For applications at very high temperatures, the KECK™ nickel-plated spring steel ground joint clamps an important addition to KECK™ conical ground joint clips made of POM (polyoxymethylene). Compared to conventional wire clamps, they are easy and safe to attach and are inherently stable.
  • Easy and safe to put on and are dimensionally stable
  • Good dimensional stability compared to conventional wire clamps
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100
Material Nickel
For Use With (Equipment) For Use at Very High Temperatures
For Use With (Application) For use at very high temperatures; Securing conical ground joints
Size NS 19