Learn More
DWK Life Sciences™ KECK™ Clip Assortment, for spherical joints, from POM
Shop All DWK Life Sciences Products
Click to view available options
For Use With (Equipment):
Secure Glass-to-Glass Joints
Unit Size:
Each
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
- For spherical joints
- Dimensionally stable with particularly good spring properties
- Easy, safe and quick to set up
- Increased safety in the laboratory as the glass surface is not scratched
- Marking of the individual sizes for better differentiation
Specifications
Specifications
| Quantity | 1 Ea. |
| Material | POM |
| Includes | 2 x KS 13, 2 x KS 19, 1 x KS 29, 1 x KS 35 |
| For Use With (Equipment) | Secure Glass-to-Glass Joints |
| For Use With (Application) | Secure glass-to-glass joints |