missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM5C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14426-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Serine/threonine kinase which acts as a negative regulator of Ras-related Rap2-mediated signal transduction to control neuronal structure and AMPA receptor trafficking. Required for normal synaptic density, dendrite complexity, as well as surface AMPA receptor expression in hippocampal neurons. Can activate the JNK and MAPK14/p38 pathways and mediates stimulation of the stress-activated protein kinase MAPK14/p38 MAPK downstream of the Raf/ERK pathway. Phosphorylates: TANC1 upon stimulation by RAP2A, MBP and SMAD1. Has an essential function in negative selection of thymocytes, perhaps by coupling NCK1 to activation of JNK1.; Isoform 4 can activate the JNK pathway. Involved in the regulation of actin cytoskeleton reorganization, cell-matrix adhesion, cell-cell adhesion and cell migration.Specifications
KDM5C | |
Polyclonal | |
Unconjugated | |
KDM5C | |
DXS1272E, Histone demethylase JARID1C, JARID1C, KDM5C, lysine (K) demethylase 5C, Lysine specific demethylase 5C, MRXJ, MRXSJ, Protein SmcX, Protein Xe169, SMCX, XE169 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20591, 317432, 8242 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41229, P41230 | |
Kdm5c | |
KDM5C Fusion Protein Ag5791 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |