Learn More
KDM1 Mouse anti-Human, Unconjugated, Clone: 2F10A12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67037-1-IG-20UL
 View more versions of this product
Product Code. 16849425
  • 1710.33 NOK / 20µL
Description
The KDM1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KDM1. This antibody recognizes human, mouse, rat antigen. The KDM1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
KDM1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
2F10A12
Unconjugated
20 μL
23028
IgG1