Description
The KCTD2/5/17 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KCTD2/5/17. This antibody recognizes human, mouse, rat antigen. The KCTD2/5/17 antibody has been validated for the following applications: FC, IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
| Antigen | KCTD2/5/17 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 333 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 54442 |
| Target Species | Human, Rat, Mouse |
| Show More |