Learn More
KCTD15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20128-1-AP-20UL
 View more versions of this product
Product Code. 16859624
  • 1710.33 NOK / 20µL
Description
Childhood and adult obesity in the United States and to a lesser extent the rest of the world has increased dramatically over the past decade. Both environmental and genetic factors are involved in the onset and progression of weight gain. Recently, the potassium channel KCTD15 was identified as a genetic loci associated with higher than normal body mass index (BMI) in humans along with genes such as GNPDA2, MTCH2, FTO, and TMEM18. Further studies on single nucleotide polymorphisms (SNPs) in non-diabetic and diabetic patients showed that FTO was most strongly associated with obesity while MTCH2 and GNPDA2 were still significantly associated with higher than normal BMI levels. At least two isoforms of KCTD15 are known to exist.
Specifications
Specifications
KCTD15
Polyclonal
Unconjugated
KCTD15
KCTD15
Rabbit
Antigen Affinity Chromatography
RUO
233107, 499129, 79047
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K0E1, Q96SI1
Kctd15
KCTD15 Fusion Protein Ag13962
20 μL
Primary
Mouse, Human, Rat