missing translation for 'onlineSavingsMsg'
Learn More

KCTD10 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16835535 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16835535 150 μL 150µL
16845535 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16835535

Brand: Proteintech 272791AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCTD10, a rat potassium channel tetramerization domain-containing 10 gene is a novel member of the polymerase delta-interacting protein 1 (PDIP1) gene family. KCTD10 shares significant similarity in amino acid sequence to PDIP1 and can interact with the small subunit of DNA polymerase delta and PCNA as PDIP1 does. Like PDIP1, the expression of KCTD10 gene can be induced by TNF-alpha in NIH3T3 cells.

Specifications

Antigen KCTD10
Applications Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KCTD10
Gene Accession No. Q9H3F6
Gene Alias KCTD10, MSTP028, ULR061, ULRO61
Gene Symbols KCTD10
Host Species Rabbit
Immunogen KCTD10 Fusion Protein Ag25971
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83892
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.