missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD10 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27279-1-AP-20UL
This item is not returnable.
View return policy
Description
KCTD10, a rat potassium channel tetramerization domain-containing 10 gene is a novel member of the polymerase delta-interacting protein 1 (PDIP1) gene family. KCTD10 shares significant similarity in amino acid sequence to PDIP1 and can interact with the small subunit of DNA polymerase delta and PCNA as PDIP1 does. Like PDIP1, the expression of KCTD10 gene can be induced by TNF-alpha in NIH3T3 cells.Specifications
| KCTD10 | |
| Polyclonal | |
| Unconjugated | |
| KCTD10 | |
| KCTD10, MSTP028, ULR061, ULRO61 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83892 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9H3F6 | |
| KCTD10 | |
| KCTD10 Fusion Protein Ag25971 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction