missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNN4 Mouse anti-Human, Mouse, Rat, Clone: 2A6G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60276-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is part of a potentially heterotetrameric voltage-independent potassium channel that is activated by intracellular calcium. Activation is followed by membrane hyperpolarization, which promotes calcium influx. The encoded protein may be part of the predomit calcium-activated potassium channel in T-lymphocytes. This gene is similar to other KCNN family potassium channel genes, but it differs enough to possibly be considered as part of a new subfamily.Specifications
KCNN4 | |
Monoclonal | |
1.05 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15554, O89109, Q9QYW1 | |
KCNN4 | |
KCNN4 Fusion Protein Ag19774 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG2a |
Western Blot | |
2A6G5 | |
Unconjugated | |
KCNN4 | |
hIKCa1, hKCa4, hSK4, IK1, IKCA1, KCa3.1, KCA4, KCNN4, Putative Gardos channel, SK4, SKCa 4, SKCa4 | |
Mouse | |
Protein A | |
RUO | |
16534, 3783, 65206 | |
-20°C | |
Liquid |