Learn More
Klever KCJ-1 Kutter
Product Code. 17788977
Change view
Click to view available options
Color:
Black
Blue
Orange
Red
Yellow
Unit Size:
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17788977 Blue Pack of 50
17798977 Orange Pack of 50
17708987 Black Pack of 50
17728987 Red Pack of 50
17748987 Yellow Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17788977 Supplier Klever Supplier No. KCJ1B
  • ONLINE EXCLUSIVE PRICE
    2039.48 NOK
    1575.99 NOK / Pack of 50
    Save 463.49 NOK
    22% Off
Estimated Shipment: 21-12-2026
KCJ-1 Klever Kutter - Recessed blade reduces cut injuries and damaged goods.
  • Recessed blade reduces cut injuries and damaged goods
  • Cuts boxes, film, tape, and thousands more applications
  • Small, convenient, and durable with high quality carbon steel blade
  • Disposable
Specifications
Blade Material Carbon Steel
Product Type Disposable Cutter
Disposable Single-use
Sterility Non-sterile
Color Blue