Learn More
KC7F2, MedChemExpress
Product Code. 16490098
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490098 5 mg 5mg
16433647 50 mg 50mg
16417638 10 mg 10mg
16407546 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16490098 Supplier Medchem Express Supplier No. HY187775mg
MedChemExpress KC7F2 is a potent hypoxia inducible factor-1 (HIF-1) pathway inhibitor with an IC50 of 20 μM in LN229-HRE-AP cells, and with potential as a cancer therapy agent.
KC7F2 is a potent hypoxia inducible factor-1 (HIF-1) pathway inhibitor with an IC50 of 20 μM in LN229-HRE-AP cells, and with potential as a cancer therapy agent.
Specifications
Chemical Name or Material KC7F2
CAS 927822-86-4
Color White
Purity Grade Notes Research
Molecular Formula C16H16Cl4N2O4S4
Quantity 5 mg
Solubility Information DMSO : ≥ 32 mg/mL (56.10 mM)
SMILES O=S(C1=CC(Cl)=CC=C1Cl)(NCCSSCCNS(C2=CC(Cl)=CC=C2Cl)(=O)=O)=O
Molecular Weight (g/mol) 570.38
Formula Weight 570.38
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease