Learn More
Nordvlies Katrin Toilet 2-Roll Dispenser
Product Code. 15700907
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15700907 Black Each
15701097 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15700907 Supplier Nordvlies Supplier No. 104452
  • ONLINE EXCLUSIVE PRICE
    838.00 NOK
    685.71 NOK / Pack of 1
    Save 152.29 NOK
    18% Off
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
The Katrin Inclusive Toilet roll dispenser is ideal for all washrooms, and is available with a wide range of conventional toilet roll solutions.
  • Practical dispenser for conventional toilet rolls
  • Holds 2 toilet rolls (including 115 mm luxury quality rolls)
  • Made from ABS Plastic Robust and reliable
  • Inspection window shows when it is time to refill the paper
  • Suitable for left and right hand corners (switchable roll holder)
  • Simple to load, install and maintain
  • Lockable
Specifications
Product Type Toilet Paper Dispenser
Color Black
Dimensions (L x W x H) 300 x 145 x 145 mm
For Use With (Application) Dispensing Toilet Paper