Learn More
Nordvlies Katrin Inclusive Gigant Toilet S Dispenser
Product Code. p-7225995
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
Product Code. Color unitSize
15701077 White Each
15781087 Black Each
2 options
Product Code. 15701077
Brand: Nordvlies 90069
Only null left
Add to basket
Add to basket
The Katrin Gigant Dispensers are particularly well-suited for areas with frequent use.
  • Dispenser for big Gigant toilet rolls for high frequency usage
  • The lock can be used with or without a key
  • The whole paper roll can be used economically with integrated stub roll holder
  • Easy paper tear-off to all sides and all edges
  • No searching around inside the dispenser opening for the loose end of the paper roll
  • Katrin Gigant S Dispensers are suitable for S sized Gigant rolls
  • New flexible roll brake for better dispensing
  • Main raw materials of major plastic parts and lock: ABS (Acrylonitrile butadiene styrene), PC (polycarbonate), POM (polyoxymethylene)
  • Katrin Inclusive dispensers are resistant to high temperatures
  • Comply with UL94 Fire Safety and Fire Protection regulations (EU)
Specifications
Product Type Dispenser
Color White
Dimensions (L x W x H) 245 x 239 x 151 mm
For Use With (Application) Dispensing Hand Towels
Size Small