Learn More
Nordvlies Katrin Inclusive Gigant Toilet S Dispenser
Product Code. 15701077
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15701077 White Each
15781087 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15701077 Supplier Nordvlies Supplier No. 90069
  • ONLINE EXCLUSIVE PRICE
    723.00 NOK
    621.26 NOK / Pack of 1
    Save 101.74 NOK
    14% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
The Katrin Gigant Dispensers are particularly well-suited for areas with frequent use.
  • Dispenser for big Gigant toilet rolls for high frequency usage
  • The lock can be used with or without a key
  • The whole paper roll can be used economically with integrated stub roll holder
  • Easy paper tear-off to all sides and all edges
  • No searching around inside the dispenser opening for the loose end of the paper roll
  • Katrin Gigant S Dispensers are suitable for S sized Gigant rolls
  • New flexible roll brake for better dispensing
  • Main raw materials of major plastic parts and lock: ABS (Acrylonitrile butadiene styrene), PC (polycarbonate), POM (polyoxymethylene)
  • Katrin Inclusive dispensers are resistant to high temperatures
  • Comply with UL94 Fire Safety and Fire Protection regulations (EU)
Specifications
Product Type Dispenser
Color White
Dimensions (L x W x H) 245 x 239 x 151 mm
For Use With (Application) Dispensing Hand Towels
Size Small