Learn More
Nordvlies Katrin Basic System Towel M
Product Code. 15701007
Change view
Click to view available options
Color:
White
Unit Size:
6 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15701007 White 6 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15701007 Supplier Nordvlies Supplier No. 46020
  • ONLINE EXCLUSIVE PRICE
    1195.00 NOK
    1025.63 NOK / 6 rolls
    Save 169.37 NOK
    14% Off
View all promotions
Estimated Shipment: 30-03-2027
Thanks to the windmill embossing, the Katrin Basic System hand towel paper is softer and even more absorbent than ever before.
  • A hygienic solution based on absorbent recycled fiber paper for effective drying
  • 1-Ply, natural white
  • Flat sheets are fed out in the right size
  • Zero wastage with all the paper on the rolled used up
  • Large amount of paper on the roll
  • When used in conjunction with the Katrin Inclusive System Towel Dispenser, the number of sheets per roll will be 805
  • Suitable for all washrooms but especially where there is medium to high traffic
  • Soft and strong
  • Controlled consumption and reduced cost in use
  • Certified under the Nordic Swan Eco Label
Specifications
Color White
Disposable Single-use
Dimensions (L x W) 180 m x 21 cm
Packaging Quantity 805 Sheets
Ply 1-ply