missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KATNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14969-1-AP-20UL
This item is not returnable.
View return policy
Description
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. Katanin is a member of the AAA family of ATPases.Specifications
| KATNB1 | |
| Polyclonal | |
| Unconjugated | |
| KATNB1 | |
| KAT, Katanin p80 subunit B1, KATNB1, p80 katanin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10300, 291852, 74187 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BG40, Q9BVA0 | |
| Katnb1 | |
| KATNB1 Fusion Protein Ag6836 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction