missing translation for 'onlineSavingsMsg'
Learn More

KATNA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16857474 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16857474 20 μL 20µL
16847474 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16857474

Brand: Proteintech 175601AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.

Specifications

Antigen KATNA1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KATNA1
Gene Accession No. O75449, Q6E0V2, Q9WV86
Gene Alias Katanin p60 subunit A1, KATNA1, p60 katanin
Gene Symbols Katna1
Host Species Rabbit
Immunogen KATNA1 Fusion Protein Ag11753
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11104, 23924, 292464
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.