Learn More
SI Analytics™ Karl Fischer Titration Vessel
Product Code. p-8012650 Shop All SI Analytics Products
Change view
Click to view available options
Volume (Metric):
30 to 150 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
10598221 30 to 150 mL Each
1 options
Product Code. 10598221
Brand: SI Analytics™ 285216677
Only null left
Add to basket
Add to basket
SI Analytics™ Karl Fischer Titration Vessel is constructed of DURAN™ glass and comes equipted with screw threads, a titration tip, and more.
  • Includes a removable head made of polypropylene/PTFE
  • Includes 1 drilling NS 19, NS 14.5, NS 7.5 and 3 drillings with screw threads
  • Includes a titration tip, moisture trap and weighing funnel
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Vessel
Includes Removable head, moisture trap, and weighing funnel
Material DURAN™ Glass
Volume (Metric) 30 to 150 mL