Learn More
Karl Fischer Aqualine™ Titrant 2, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Product Code. p-7073247 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
Product Code. 10786291
Brand: Fisher Chemical K/2150/15
Request Bulk or Custom Format
Chemical Identifiers
MDL Number 132730
Specifications
Boiling Point 64°C
Color Black to Red
Physical Form Liquid
Quantity 1 L