Learn More
Thermo Scientific Chemicals Kallikrein Inhibitor
Product Code. 15457509
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Product Code. 15457509
Brand: Thermo Scientific Chemicals J66234.M^
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 97145-43-2
Molecular Formula C35H55N11O9
Synonym N-Acetyl-Pro-Phe-Arg-Ser-Val-Gln-NH{2}; KKI 5
Specifications
Quantity 1 mg
Description White to off-white
Content And Storage -30°C to -10°C
Formula Weight 773.89
Solubility Soluble in water at 1mg/ml
Product Type Kallikrein Inhibitor
Form Solid