missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10514-2-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.Specifications
Kallikrein 5 | |
Polyclonal | |
Unconjugated | |
KLK5 | |
Kallikrein 5, Kallikrein like protein 2, kallikrein related peptidase 5, KLK L2, KLK5, KLKL2, SCTE, Stratum corneum tryptic enzyme | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25818 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y337 | |
KLK5 | |
Kallikrein 5 Fusion Protein Ag0799 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |