Learn More
Scharlab KALIUMHYDROXID PURISS 1KGSegregation: Separated from acids.
Brand:  Scharlab PO02661000
KALIUMHYDROXID PURISS 1KGSegregation: Separated from acids.
Product Code. 11304244
  • ONLINE EXCLUSIVE PRICE
    541.80 NOK
    385.62 NOK / 1kg
    Save 156.18 NOK
    28% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Potassium hydroxide, pellets, extra pure, Pharmpur, Ph Eur, BP, NF
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .