Learn More
Robin™ Kahn Complete Shaker
Product Code. 11959617
Change view
Click to view available options
Speed Range:
100 rpm
180 rpm
40 to 120 rpm
Unit Size:
Each
Product Code. Speed Range unitSize
11959617 180 rpm Each
11959607 100 rpm Each
11919607 40 to 120 rpm Each
3 options
Product Code. 11959617
Brand: Robin™ PB008
Only null left
Add to basket
Add to basket
Versatile shaker for mixing or agitating ingredients and solutions in flasks, bottles, beakers, and Kahn racks. Robin™ Kahn Complete Shaker is available in different shaker speed ranges.
  • Mixes or agitates ingredients and solutions in a variety of containers including, but not limited to, the following:
    • Flasks
    • Bottles
    • Beakers
    • Kahn racks
Specifications
Load Bearing Capacity (Metric) 6.5 kg
Speed Range 180 rpm
Electrical Requirements 230 V, 50 Hz, 25 W
Wattage 25 W
Product Type Kahn Complete Shaker
Frequency 50 Hz
Depth (Metric) 250 mm
Dimensions (D x W x H) Interior 250 x 360 x 130 mm
Height (Metric) 130 mm
Width (Metric) 360 mm
Timer Range 60min.