Learn More
Robin™ Kahn Complete Shaker
Product Code. 11959607
Change view
Click to view available options
Speed Range:
100 rpm
180 rpm
40 to 120 rpm
Unit Size:
Each
Product Code. Speed Range unitSize
11959607 100 rpm Each
11959617 180 rpm Each
11919607 40 to 120 rpm Each
3 options
Product Code. 11959607
Brand: Robin™ PB007
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Versatile shaker for mixing or agitating ingredients and solutions in flasks, bottles, beakers, and Kahn racks. Robin™ Kahn Complete Shaker is available in different shaker speed ranges.
  • Mixes or agitates ingredients and solutions in a variety of containers including, but not limited to, the following:
    • Flasks
    • Bottles
    • Beakers
    • Kahn racks
Specifications
Load Bearing Capacity (Metric) 6.5 kg
Speed Range 100 rpm
Electrical Requirements 230 V, 50 Hz, 25 W
Wattage 25 W
Product Type Kahn Complete Shaker
Frequency 50 Hz
Depth (Metric) 250 mm
Dimensions (D x W x H) Interior 250 x 360 x 130 mm
Height (Metric) 130 mm
Width (Metric) 360 mm
Timer Range 15min.