Learn More
HUBER K20 Cooling Bath
K20 Refrigerant bath, with natural refrigerant
Brand:  HUBER 2011.0022.00
Additional Details : Weight : 30.00000kg
Product Code. 10276473
  • 55066.36 NOK / Each
Estimated Shipment: 23-07-2024
Description
K20 Refrigerant bath, with antural refrigerant.
  • Temperature range: -30°C to 200°C
  • Cooling capacity (0°C): 0.35kW
  • Power supply: 230V 1∼ 50/60Hz
Specifications
Specifications
20°C at 0.4 kW, 0°C at 0.35 kW, -10°C at 0.27°CkW, to 20°C at 0.16°CkW, to 30°C (at 0.06)
K20
350 mm
30 kg
50/60 Hz
-30°C to +200°C
3 years for removable (Plug & Play) electronic controls, 2 years for refrigeration components, 1 year for mechanical and electrical components
450 mm
230 V