Learn More
JW 642, R&D Systems™
Product Code. 30002020
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30002020 50 mg 50mg
30002006 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30002020 Supplier Tocris Bioscience Supplier No. 4906/50
 Restricted Item
Estimated Shipment: 27-11-2026
A Tocris product, part of R&D Systems. MAGL inhibitor
Potent and selective monoacylglycerol lipase (MAGL) inhibitor (IC50 = 3.7 nM). Displays >1000-fold selectivity for MAGL over fatty acid amide hydrolase (IC50 = 20.6 μM). Analog of JZL 195 (Cat. No. 4715).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1416133-89-5
Molecular Formula C21H20F6N2O3
Molecular Weight (g/mol) 462.392
InChI Key AVSCNEOUWSVZEY-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoropropan-2-yl 4-3-phenoxybenzyl piperazine-1-carboxylate, 1,1,1,3,3,3-hexafluoropropan-2-yl 4-3-phenoxyphenyl methyl piperazine-1-carboxylate, 4-3-phenoxyphenyl methyl-1-piperazinecarboxylic acid 2,2,2-trifluoro-1-trifluoromethyl ethyl ester
PubChem CID 71656520
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate
SMILES C1CN(CCN1CC2=CC(=CC=C2)OC3=CC=CC=C3)C(=O)OC(C(F)(F)F)C(F)(F)F
Specifications
Color White
Quantity 50 mg
Formula Weight 462.39
Percent Purity >98%
Physical Form Solid
Product Line Tocris
Chemical Name or Material JW 642