Learn More
Justrite™ Cease-Fire™ Head
Convert standard metal drum into fire-safe waste receptacle
1255.62 NOK
Specifications
Certifications/Compliance | FM, UL |
---|---|
Quantity | 1 Ea. |
For Use With (Application) | Install easily on existing metal drums |
Material | Aluminum |
Product Type | Cease-Fire Head |
Product Code | Brand | Opening Size (English) | Diameter (English) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Opening Size (English) | Diameter (English) Outer | Price | Quantity & Availability | ||||
11973669
|
Justrite™
26555 |
7.8 in. | 24 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Install easily on existing metal drums
- Aluminum construction is virtually corrosion free for easy clean-up
Certifications
FM approved
Specifications
FM, UL | |
Install easily on existing metal drums | |
Cease-Fire Head |
1 Ea. | |
Aluminum |