Learn More
Justrite™ Accessory Shelf
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
17 gal. Piggyback, All 2-door 30, 40 and 45 gal. Safety Cabinets
missing translation for 'unitSize'
Each
Description
- Made of polyethylene
- Curved edge prevents leakage and spills
- Fits Shelf Model No 29937 for 17 gal. Piggyback and all 2 door 30, 40 and 45 gal. cabinets
Specifications
Specifications
| For Use With (Equipment) | 17 gal. Piggyback, All 2-door 30, 40 and 45 gal. Safety Cabinets |
| Includes | Slightly curved edge |
| Weight (English) | 3 lb. |
| Weight (Metric) | 1.3 kg |
| Width (Metric) Exterior | 34.3 cm |
| Material | Polyethylene |
| Product Type | Shelf Tray |
| Width (English) Exterior | 13.5 in. |