Learn More
3M™ Jupiter™ Air-Fed Powered Turbo Respirators
Click to view available options
Battery Life:
4 to 8 hours
Unit Size:
Each
Description
- Decontamination comfort belt for improved fit and easier cleaning
- Option of five filter types to select from, depending on work environment
- Modern, ergonomic design
- Can be worn during shower decontamination
- Fully compatible with all 3M™ Versaflo™ Headtops and Breathing Tubes
- Integrated acoustic and visual alarm indicates if airflow to the headtop is reduced or battery charge runs low
Specifications
Specifications
| Battery Life | 4 to 8 hours |
| Battery Type | NiMH |
| Certifications (CSA) | CE Approved |
| Certifications (EN) | EN 12941 TH2, EN 12941 TH3 |
| Color | Blue, Black |
| Product Type | PAPR Assembly, No Headtop |
| For Use With (Application) | Used to Provides Protection Against Particulates, Gas and Vapors |
| Includes | Turbo unit only: motor, fan, comfort belt and instructions; helmet and hose not included |
| Model | 0850010P |
| Quantity | 1 Ea. |