Learn More
Trajan™ Jumbo Syringe
Description
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
- Designed for holding and dispensing large volumes of gas
- Heavy duty acrylic barrels
- Easy access to sample for the addition of standards or removal of subsample via secondary port
- Plunger stem can be unscrewed for ease of transportation and storage
- Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Specifications
| Material | Acrylic, Acrylic (Barrel) |
| Diameter (Metric) Outer | 100 mm |
| For Use With (Application) | For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling |
| Length (Metric) | 159.1 mm |
| Product Type | Jumbo Syringe |
| Model | 1000MAR-LL-GT |
| Volume (Metric) | 1000 mL |
| Product Line | Trajan |