Learn More
Trajan™ Jumbo Syringe
Product Code. 10045470
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
2000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10045470 2000 mL Each
10751042 1000 mL Each
10300811 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10045470 Supplier Trajan™ Supplier No. 009930
Only null left
Add to basket
Add to basket
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
  • Designed for holding and dispensing large volumes of gas
  • Heavy duty acrylic barrels
  • Easy access to sample for the addition of standards or removal of subsample via secondary port
  • Plunger stem can be unscrewed for ease of transportation and storage
  • Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Material Acrylic, Acrylic (Barrel)
Diameter (Metric) Outer 130 mm barrel
For Use With (Application) For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling
Length (Metric) 245 mm
Product Type Jumbo Syringe
Graduations 200 mL major scale, 100 mL minor scale
Model 2000MAR-LL-GT
Volume (Metric) 2000 mL
Product Line Trajan
Graduated Yes