missing translation for 'onlineSavingsMsg'
Learn More

JPH4 Antibody [HRP], Novus Biologicals Biologicals™

Product Code. 30499729 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30499729 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30499729 Supplier Novus Biologicals Supplier No. NBP327876H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

JPH4 Polyclonal antibody specifically detects JPH4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen JPH4
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias JP4, JP-4, JPHL1, Junctophilin 4, Junctophilin Like 1, junctophilin-4, Junctophilin-Like 1 Protein, KIAA1831
Host Species Rabbit
Immunogen JPH4 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human JPH4. The immunogen is located within amino acids 470 - 520 of JPH4.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84502
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.