missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK1 Monoclonal Antibody (OTI10D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
2832.96 NOK - 6331.68 NOK
Specifications
| Antigen | JNK1 |
|---|---|
| Clone | OTI10D8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17538124
|
Origene Technologies
TA813292S |
30 μL |
2832.96 NOK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17518124
|
Origene Technologies
TA813292 |
100 μL |
6331.68 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JNK1 Monoclonal antibody specifically detects JNK1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| JNK1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| AI849689; c-Jun N-terminal kinase 1; FLJ12099; FLJ33785; JNK; JNK1; JNK1 beta1 protein kinase; jnk1a-1; JNK1A2; JNK21B1/2; JNK3; JNK3A; JNK-46; JUN N-terminal kinase; MAP kinase 10; MAP kinase 8; MAPK 10; MAPK 8; MAPK10; Mapk8; mapk8 protein; mapk8b; MGC50974; mitogen activated protein kinase 8; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 8b; MK10; OTTHUMP00000019552; p493F12; p54 gamma; p54bSAPK; Phospho-JNK; PRKM10; PRKM8; protein kinase JNK1; protein kinase mitogen-activated 8; SAPK gamma; SAPK1; SAPK1C; stress-activated protein kinase 1; stress-activated protein kinase 1c; Stress-activated protein kinase JNK1; zgc:112379 | |
| Human recombinant protein fragment corresponding to amino acids 39-402 of human JNK1 (NP_002744) produced in E.coli. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| MAPK8 |
| OTI10D8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P45983, P49185, Q91Y86 | |
| 116554, 26419, 5599 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title