Learn More
JNJ-42165279, MedChemExpress
Product Code. 16450298
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16450298
Brand: Medchem Express HY19636
MedChemExpress JNJ-42165279 is a FAAH inhibitor with IC50 of 70 ± 8 nM and 313 ± 28 nM for hFAAH and rFAAH, respectively.
JNJ-42165279 is a FAAH inhibitor with IC50 of 70 ± 8 nM and 313 ± 28 nM for hFAAH and rFAAH, respectively.
Specifications
Chemical Name or Material JNJ-42165279
CAS 1346528-50-4
Color White
Purity Grade Notes Research
Molecular Formula C18H17ClF2N4O3
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (243.43 mM)
SMILES O=C(N1CCN(CC2=CC=C(OC(F)(F)O3)C3=C2)CC1)NC4=C(Cl)C=CN=C4
Molecular Weight (g/mol) 410.8
Formula Weight 410.8
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.