missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
JMJD1B Polyclonal antibody specifically detects JMJD1B in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | JMJD1B |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C5orf7, EC 1.14.11, EC 1.14.11.-, JHDM2B, JmjC domain-containing histone demethylation protein 2B, JMJD1B, jumonji domain containing 1B, Jumonji domain-containing protein 1B, KIAA1082chromosome 5 open reading frame 7, lysine (K)-specific demethylase 3B, lysine-specific demethylase 3B, NET22,5qNCA, Nuclear protein 5qNCA |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 400-600 of human JMJD1B (NP_057688.2).,, Sequence:, GAENKEAGKTLEQVGQGIVASAAVVTTASSTPNTVRISDTGLAAGTVPEKQKGSRSQASGENSRNSILASSGFGAPLPSSSQPLTFGSGRSQSNGVLATENKPLGFSFGCSSAQEAQKDTDLSKNLFFQCMSQTLPTSNYFTTVSESLADDSSSRDSFKQSLESLSSGLCKGRSVLGTDTKPGSKAGSSVDRKVPAESMPT |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?