missing translation for 'onlineSavingsMsg'
Learn More

JMJD1B Antibody [Alexa Fluor« 532], R&D Systems™

Product Code. 30498352 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30498352 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498352 Supplier Novus Biologicals Supplier No. NBP337990AF532
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

JMJD1B Polyclonal antibody specifically detects JMJD1B in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen JMJD1B
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias C5orf7, EC 1.14.11, EC 1.14.11.-, JHDM2B, JmjC domain-containing histone demethylation protein 2B, JMJD1B, jumonji domain containing 1B, Jumonji domain-containing protein 1B, KIAA1082chromosome 5 open reading frame 7, lysine (K)-specific demethylase 3B, lysine-specific demethylase 3B, NET22,5qNCA, Nuclear protein 5qNCA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-600 of human JMJD1B (NP_057688.2).,, Sequence:, GAENKEAGKTLEQVGQGIVASAAVVTTASSTPNTVRISDTGLAAGTVPEKQKGSRSQASGENSRNSILASSGFGAPLPSSSQPLTFGSGRSQSNGVLATENKPLGFSFGCSSAQEAQKDTDLSKNLFFQCMSQTLPTSNYFTTVSESLADDSSSRDSFKQSLESLSSGLCKGRSVLGTDTKPGSKAGSSVDRKVPAESMPT
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51780
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.