missing translation for 'onlineSavingsMsg'
Learn More

JJKK 048, MedChemExpress

Product Code. p-200096221
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. Quantity unitSize
16496927 5 mg 5mg
16452996 25 mg 25mg
16404218 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 16496927

Brand: Medchem Express HY108613

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JJKK 048 is an ultrapotent and highly selective inhibitor of Monoacylglycerol lipase (MAGL).

JJKK 048 is an ultrapotent and highly selective inhibitor of Monoacylglycerol lipase (MAGL).

Specifications

Chemical Name or Material JJKK 048
CAS 1515855-97-6
Color White
Purity Grade Notes Research
Molecular Formula C23H22N4O5
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (76.72 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N1CCC(C(C2=CC=C(OCO3)C3=C2)C4=CC=C(OCO5)C5=C4)CC1)N6N=CN=C6
Molecular Weight (g/mol) 434.44
Formula Weight 434.44
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.