Learn More
Bibby Scientific™ Jenway™ Plastic Cuvettes For UV and Visible Region Wavelength
Product Code. 11430559
Change view
Click to view available options
Capacity (Metric):
0.07 to 1.5 mL
1.5 to 3.0 mL
Unit Size:
Pack of 100
Product Code. Capacity (Metric) unitSize
11430559 1.5 to 3.0 mL Pack of 100
11475009 0.07 to 1.5 mL Pack of 100
2 options
Product Code. 11430559
Brand: Bibby Scientific™ 8307034
Only null left
Add to basket
Add to basket
Save cleaning time and avoid cross-contamination with disposable plastic cuvettes. Bibby Scientific™ Jenway™ Plastic Cuvettes For UV and Visible Region Wavelength come in a pack of 100.
  • Efficient and time saving
  • Prevents cross-contamination
Specifications
Type Cuvette
Capacity (Metric) 1.5 to 3.0 mL
Material Plastic
Quantity 100/Pk.
Path Length 10 mm
For Use With (Application) Spectral Measurement of Liquids