Learn More
Jenway™ pH Electrodes
Product Code. 11490619
Click to view available options
Length (Metric):
40 mm
90 mm
120 mm
150 mm
Connector Types:
2mm plug
3150/3205 waterproof connector
BNC
BNC/8 pin mini-DIN
S7
Diameter (Metric):
4.5 mm
6 mm
12 mm
Material:
Epoxy
Glass
Unit Size:
Each
Product Code. 11490619
Brand: Jenway™ 9026917
Only null left
Add to basket
Add to basket
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Connector Types BNC
Diameter (Metric) 6 mm
Type Refillable Spear Type Combination with Bulb End
Dimensions (L x W x H) 6 dia. x 40 mm L
For Use With (Application) measurements in small volumes and soil and slurry samples
Length (Metric) 40 mm
Material Glass
pH Range 0 to 14
Temperature (Metric) Range 0°C to 80°C