Learn More
Jenway™ pH Electrodes
Product Code. 11449489
Change view
Click to view available options
Length (Metric):
40 mm
90 mm
120 mm
150 mm
Connector Types:
2mm plug
3150/3205 waterproof connector
BNC
BNC/8 pin mini-DIN
S7
Diameter (Metric):
4.5 mm
6 mm
12 mm
Material:
Epoxy
Glass
Unit Size:
Each
Product Code. Length (Metric) Connector Types Diameter (Metric) Material unitSize
11449489 120 mm S7 12 mm Epoxy Each
13107184 150 mm BNC 6 mm Glass Each
11410499 90 mm BNC 4.5 mm Glass Each
11440499 90 mm BNC 6 mm Glass Each
11440629 120 mm 2mm plug 12 mm Glass Each
13107424 120 mm 3150/3205 waterproof connector 12 mm Epoxy Each
11490619 40 mm BNC 6 mm Glass Each
11460499 120 mm BNC/8 pin mini-DIN 12 mm Epoxy Each
11430629 120 mm BNC 12 mm Glass Each
9 options
Product Code. 11449489
Brand: Jenway™ 6800640
Only null left
Add to basket
Add to basket
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Connector Types S7
Diameter (Metric) 12 mm
Type Sealed Combination with Bulb End and Protective Cover
Dimensions (L x W x H) 12 dia. x 120 mm L
For Use With (Application) standard measurements of general solutions, field measurements or in education
Length (Metric) 120 mm
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Range 0°C to 60°C