Learn More
Jenway™ Model 731001 Visible Single Beam Spectrophotometer
Product Code. 13101744
Change view
Click to view available options
Type:
Visible Spectrophotometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
13101744 Visible Spectrophotometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13101744 Supplier Jenway™ Supplier No. 8305817
  • ONLINE EXCLUSIVE PRICE
    43725.00 NOK
    39285.41 NOK / Pack of 1
    Save 4439.59 NOK
    10% Off
View all promotions
Estimated Shipment: 08-12-2026
Ideal for routine testing in clinical, veterinary, pharmaceutical and QC laboratories.
The 73 series spectrophotometers use icon-driven software and have an improved navigation system for easy and intuitive usability. The 73 series range is comprised of four models: Models 7300 and 7305 are basic non-scanning spectrophotometers with measurement modes for absorbance, % transmittance and concentration. This model 7310 and model 7315 are advanced scanning spectrophotometers which have additional measurement modes for spectrum scanning, kinetics and quantitation. The 7310 and 7315 have the additional feature of an easy-access USB port which enables results and methods to be saved directly to USB memory stick.
  • Icon-driven software
  • Small footprint
  • Autologging capabilities
  • Save methods and results to USB memory stick
  • Three year warranty
  • Extensive range of accessories available The 73 series spectrophotometers are ideal for routine testing in clinical, veterinary, pharmaceutical and QC laboratories.
Specifications
Concentration –300 to 9999
Depth (Metric) 400 mm
Light Source Tungsten Halogen
Spectral Bandwidth 5 nm
Type Visible Spectrophotometer
Wavelength Range 320 to 1000 nm
Height (Metric) 220 mm
Optical Design Single Beam
Resolution 1 nm
Warranty 3 Years
Wavelength Accuracy ±2 nm
Weight (Metric) 6 kg
Width (Metric) 275 mm