Learn More
Cole-Parmer™ Jenway™ Cuvette Holders
Product Code. p-8012857
Change view
Click to view available options
For Use With (Equipment):
63/73/ 67 Series Spec
6850 Series Spectrophotometers
Pathlength:
10 to 100 mm
10 x 10 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Pathlength unitSize
11480539 63/73/ 67 Series Spec 10 to 100 mm Each
11454939 63/73/ 67 Series Spec 10 x 10 mm Each
13266389 6850 Series Spectrophotometers 10 to 100 mm Each
13246389 6850 Series Spectrophotometers 10 x 10 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11480539 Supplier Cole-Parmer™ Supplier No. 8305680