Learn More
JDTic dihydrochloride, MedChemExpress
Product Code. 16493998
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16493998
Brand: Medchem Express HY10487
MedChemExpress JDTic (dihydrochloride) is a potent antagonist of kappa-opioid receptors (KOR), blocking the κ-agonist U50, 488-induced antinociception.
JDTic (dihydrochloride) is a potent antagonist of kappa-opioid receptors (KOR), blocking the κ-agonist U50, 488-induced antinociception.
Specifications
Chemical Name or Material JDTic dihydrochloride
CAS 785835-79-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H41Cl2N3O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (185.68 mM; Need ultrasonic) ∣H2O : 50 mg/mL (92.84 mM; Need ultrasonic)
SMILES O=C([C@@H]1NCC2=C(C=CC(O)=C2)C1)N[C@@H](C(C)C)CN3C[C@H](C)[C@](C)(C4=CC=CC(O)=C4)CC3.[H]Cl.[H]Cl
Molecular Weight (g/mol) 538.55
Formula Weight 538.55
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.