Learn More
Cole-Parmer™ JD Honigberg™ Ice-Pak Cold Packs
Product Code. 11711029 Shop All Cole-Parmer Products
Change view
Click to view available options
Dimensions (L x W x H):
177.8 x 95.25 x 38.1 mm
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11711029 177.8 x 95.25 x 38.1 mm Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11711029 Supplier Cole-Parmer™ Supplier No. 0634520
  • ONLINE EXCLUSIVE PRICE
    1330.00 NOK
    1255.32 NOK / Pack of 24
    Save 74.68 NOK
    5% Off
Estimated Shipment: 25-01-2027
Specifications
Quantity 24 Pk.
Product Type Cold Pack
For Use With (Application) Refrigerate Samples For Shipping
Dimensions (L x W x H) 177.8 x 95.25 x 38.1 mm
Temperature (Metric) Operating -6°C