Learn More
Jasplakinolide, TRC
Product Code. 30405131
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405131 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30405131 Supplier Toronto Research Chemicals Supplier No. TRCJ21070050UG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Jasplakinolide
CAS 102396-24-7
Purity Grade Notes HPLC
Molecular Formula C36H45BrN4O6
Quantity 50 μg
Synonym NSC 613009, [2S-[1(S*),2R*,4E,6S*,8R*]]-N-[2-Bromo-N-[N-(8-hydroxy-2,4,6-trimethyl-1-oxo-4-nonenyl)-L-alanyl]-N-methyl-D-tryptophyl]-3-(4-hydroxyphenyl)-β-alanine ρ-Lactone, 1-Oxa-5,8,11-triazacyclononadecane β-alanine deriv., (+)-Jasplakinolide, Jaspamide
InChI Formula InChI=1S/C36H45BrN4O6/c1-20-15-21(2)17-23(4)47-32(43)19-30(25-11-13-26(42)14-12-25)40-35(45)31(18-28-27-9-7-8-10-29(27)39-33(28)37)41(6)36(46)24(5)38-34(44)22(3)16-20/h7-15,21-24,30-31,39,42H,16-19H2,1-6H3,(H,38,44)(H,40,45)/b20-15+/t21-,22-,23-,24-,30+,31+/m0/s1
SMILES C[C@H]1C[C@@H](C)\C=C(/C)\C[C@H](C)C(=O)N[C@@H](C)C(=O)N(C)[C@H](Cc2c(Br)[nH]c3ccccc23)C(=O)N[C@H](CC(=O)O1)c4ccc(O)cc4
IUPAC Name (4R,7R,10S,13S,15E,17R,19S)-7-[(2-bromo-1H-indol-3-yl)methyl]-4-(4-hydroxyphenyl)-8,10,13,15,17,19-hexamethyl-1-oxa-5,8,11-triazacyclononadec-15-ene-2,6,9,12-tetrone
Molecular Weight (g/mol) 709.67
Formula Weight 708.2522
Percent Purity >95