missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human JAK1, GST Tag Recombinant Protein
Janus kinase 1 (JAK1) is a protein tyrosine kinase.
Brand: Invitrogen™ PV4774
Additional Details : Weight : 0.00010kg
Description
Janus kinase 1 (JAK1), is a protein tyrosine kinase that is involved in the interferon-alpha⁄beta and -gamma signal transduction pathways.Specifications
NP_002218.2 | |
3716 | |
10 μg | |
JAK Kinase Family | |
Human | |
GST Tag | |
Biochemical terbium phospho-specific activity, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
JAK1 | |
Baculovirus | |
Recombinant, Cytoplasmic, Catalytic | |
Cytoplasmic Tyrosine Kinases |
Kinase Assay | |
60.7 kDa | |
JAK1, JAK1A, JAK1B | |
aa 866-1154 | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™ | |
Janus kinase 1 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |